ENA024295078 Enamine

1-(1,1-dioxidotetrahydrothien-3-yl)-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD05668167
  • Cas Number : 27280-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215...217
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09040

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09040
Availability details for
Quantity
  • Availability
  • Add to Cart