ENA024295082 Enamine

3-(1-hydroxy-1H-1,2,3-benzotriazole-6-sulfonamido)benzoic acid

  • MDL number MFCD06336107
  • Cas Number : 749906-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09044
Availability details for
Quantity
  • Availability
  • Add to Cart