ENA024295090 Enamine

2-chloro-N-{[(4-fluorophenyl)carbamoyl]methyl}-N-methylacetamide

  • MDL number MFCD05262717
  • Cas Number : 743440-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09052
Availability details for
Quantity
  • Availability
  • Add to Cart