ENA024295094 Enamine

2-(4-cyanobenzenesulfonamido)acetic acid

  • MDL number MFCD05262724
  • Cas Number : 214841-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09056
Availability details for
Quantity
  • Availability
  • Add to Cart