ENA024295096 Enamine

2-{[(3,5-dimethyl-1,2-oxazol-4-yl)methyl]sulfanyl}acetic acid

  • MDL number MFCD03011609
  • Cas Number : 446830-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09058
Availability details for
Quantity
  • Availability
  • Add to Cart