ENA024295101 Enamine

N,N-diethyl-3-nitro-4-(piperazin-1-yl)benzene-1-sulfonamide

  • MDL number MFCD05262728
  • Cas Number : 743440-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-72
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09063
Availability details for
Quantity
  • Availability
  • Add to Cart