ENA024295106 Enamine

4-(chloromethyl)-6-ethyl-7-hydroxy-2H-chromen-2-one

  • MDL number MFCD05263744
  • Cas Number : 743452-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09068

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09068
Availability details for
Quantity
  • Availability
  • Add to Cart