ENA024295180 Enamine

1-chloro-4-[(2-chloroethyl)sulfanyl]benzene

  • MDL number MFCD00018965
  • Cas Number : 14366-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-31
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09143
Availability details for
Quantity
  • Availability
  • Add to Cart