ENA024295181 Enamine

1,4-dichloro-2-[(2-chloroethyl)sulfanyl]benzene

  • MDL number MFCD06655029
  • Cas Number : 742104-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09144

Safety Information

signalword Danger
pcodes P201,P202,P260,P261,P264,P271,P280,P281,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P308+P313,P310,P311,P312,P361,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-09144
pictograms GHS05,GHS06,GHS08
hstatements H311,H314,H331,H350
Availability details for
Quantity
  • Availability
  • Add to Cart