ENA024295198 Enamine

4,8-dimethylquinoline-2-thiol

  • MDL number MFCD06336109
  • Cas Number : 53761-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09161

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09161
Availability details for
Quantity
  • Availability
  • Add to Cart