ENA024295199 Enamine

6-chloro-4-(chloromethyl)-7-ethyl-2H-chromen-2-one

  • MDL number MFCD05263784
  • Cas Number : 743452-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09162

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09162
Availability details for
Quantity
  • Availability
  • Add to Cart