ENA024295208 Enamine

2-chloro-1-[1-(2-furylmethyl)-2,5-dimethyl-1H-pyrrol-3-yl]propan-1-one

  • MDL number MFCD05263812
  • Cas Number : 743453-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09171

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09171
Availability details for
Quantity
  • Availability
  • Add to Cart