ENA024295210 Enamine

2-(4-fluorophenyl)-2-(morpholin-4-yl)ethan-1-amine

  • MDL number MFCD06655030
  • Cas Number : 851169-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09173
Availability details for
Quantity
  • Availability
  • Add to Cart