ENA024295211 Enamine

2-(2-methanesulfonyl-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)acetohydrazide

  • MDL number MFCD06336104
  • Cas Number : 100879-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09174
Availability details for
Quantity
  • Availability
  • Add to Cart