ENA024295221 Enamine

2-[(3-bromophenyl)methoxy]benzoic acid

  • MDL number MFCD05263818
  • Cas Number : 743453-43-2
  • Molecular Weight : 307.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-09184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09184
Availability details for
Quantity
  • Availability
  • Add to Cart