ENA024295226 Enamine

3-{[(4-methoxy-3-nitrophenyl)sulfonyl]amino}propanoic acid

  • MDL number MFCD04639673
  • Cas Number : 741731-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09189

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09189
Availability details for
Quantity
  • Availability
  • Add to Cart