ENA024295239 Enamine

2-({5-[(3,5-dimethylphenyl)amino]-1,3,4-thiadiazol-2-yl}sulfanyl)propanoic acid

  • MDL number MFCD05264093
  • Cas Number : 743457-06-9
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-128
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-09202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09202
Availability details for
Quantity
  • Availability
  • Add to Cart