ENA024295248 Enamine

2-hydrazinyl-4,8-dimethylquinoline

  • MDL number MFCD02197923
  • Cas Number : 57369-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09211
Availability details for
Quantity
  • Availability
  • Add to Cart