ENA024295262 Enamine

N-benzyl-6-fluoro-1,3-benzothiazol-2-amine

  • MDL number MFCD05266694
  • Cas Number : 131169-64-7
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09225
Availability details for
Quantity
  • Availability
  • Add to Cart