ENA024295270 Enamine

11-tert-butyl-4-(2,6-dimethylphenyl)-5-sulfanyl-8-thia-4,6-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),5-trien-3-one

  • MDL number MFCD06334594
  • Cas Number : 744227-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09233
Availability details for
Quantity
  • Availability
  • Add to Cart