ENA024295294 Enamine

N-(1-cyclopropylethyl)-3-(trifluoromethyl)aniline

  • MDL number MFCD06660706
  • Cas Number : 885460-34-4
  • Molecular Weight : 229.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(1-cyclopropylethyl)-3-(trifluoromethyl)aniline
Inchi Code 1S/C12H14F3N/c1-8(9-5-6-9)16-11-4-2-3-10(7-11)12(13,14)15/h2-4,7-9,16H,5-6H2,1H3
Inchi Key KBXWRPZHSHUJTO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-09257

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09257
Availability details for
Quantity
  • Availability
  • Add to Cart