ENA024295295 Enamine

{allyl[(4-chlorophenyl)sulfonyl]amino}acetic acid

  • MDL number MFCD05265118
  • Cas Number : 744203-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136...138
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09258

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09258
Availability details for
Quantity
  • Availability
  • Add to Cart