ENA024295303 Enamine

3,5-dimethylisoxazole-4-carbaldehyde

  • MDL number MFCD02681977
  • Cas Number : 54593-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09266

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-09266
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart