ENA024295305 Enamine

1-butyl-6-oxo-1,6-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD05263553
  • Cas Number : 103854-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09268
Availability details for
Quantity
  • Availability
  • Add to Cart