ENA024295307 Enamine

5-chloro-1-(2,4-dimethylphenyl)-3-methyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD05263556
  • Cas Number : 956201-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09270

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09270
Availability details for
Quantity
  • Availability
  • Add to Cart