ENA024295319 Enamine

2-(chloromethyl)-1-ethyl-N,N-dimethyl-1H-benzimidazole-5-sulfonamide

  • MDL number MFCD04621664
  • Cas Number : 730951-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09282
Availability details for
Quantity
  • Availability
  • Add to Cart