ENA024295320 Enamine

3-{1-butyl-5-[(dimethylamino)sulfonyl]-1H-benzimidazol-2-yl}propanoic acid

  • MDL number MFCD05267695
  • Cas Number : 744242-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09283

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09283
Availability details for
Quantity
  • Availability
  • Add to Cart