ENA024295347 Enamine

6-fluoro-2,3-dihydrospiro[1lambda6-benzothiopyran-4,4'-imidazolidine]-1,1,2',5'-tetrone

  • MDL number MFCD05267731
  • Cas Number : 66892-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-251
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09310
Availability details for
Quantity
  • Availability
  • Add to Cart