ENA024295349 Enamine

2-{[bis(4-fluorophenyl)methyl]sulfanyl}acetic acid

  • MDL number MFCD05270835
  • Cas Number : 90212-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09312
Availability details for
Quantity
  • Availability
  • Add to Cart