ENA024295353 Enamine

(2E)-4-{4-[(3-nitrophenyl)sulfonyl]piperazin-1-yl}-4-oxobut-2-enoic acid

  • MDL number MFCD05267056
  • Cas Number : 744230-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09316

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09316
Availability details for
Quantity
  • Availability
  • Add to Cart