ENA024295362 Enamine

2-(1H-1,2,3,4-tetrazol-1-yl)benzoic acid

  • MDL number MFCD00657953
  • Cas Number : 116570-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194-195
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09325
Availability details for
Quantity
  • Availability
  • Add to Cart