ENA024295369 Enamine

(2-oxo-1,3-benzoxazol-3(2H)-yl)acetic acid

  • MDL number MFCD01547452
  • Cas Number : 13610-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09332

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09332
Availability details for
Quantity
  • Availability
  • Add to Cart