ENA024295371 Enamine

6-oxo-1-propyl-1,6-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD05263570
  • Cas Number : 171672-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09334

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09334
Availability details for
Quantity
  • Availability
  • Add to Cart