ENA024295372 Enamine

2-chloro-N-{[(2,3,4-trifluorophenyl)carbamoyl]methyl}acetamide

  • MDL number MFCD05267735
  • Cas Number : 744243-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09335
Availability details for
Quantity
  • Availability
  • Add to Cart