ENA024295373 Enamine

2-(4-formyl-2-methoxyphenoxy)acetamide

  • MDL number MFCD01050452
  • Cas Number : 186685-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09336
Availability details for
Quantity
  • Availability
  • Add to Cart