ENA024295381 Enamine

2-chloro-1-[2,5-dimethyl-1-(2,2,2-trifluoroethyl)-1H-pyrrol-3-yl]ethan-1-one

  • MDL number MFCD05270015
  • Cas Number : 746630-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09345
Availability details for
Quantity
  • Availability
  • Add to Cart