ENA024295400 Enamine

2-[(1,1-dioxo-1lambda6,2-benzothiazol-3-yl)amino]-3-methylbutanoic acid

  • MDL number MFCD05270831
  • Cas Number : 1396968-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234-235
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09364
Availability details for
Quantity
  • Availability
  • Add to Cart