ENA024295403 Enamine

5-(pyrrolidin-1-ylmethyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD06357416
  • Cas Number : 790263-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228...230
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09367
Availability details for
Quantity
  • Availability
  • Add to Cart