ENA024295409 Enamine

4-(4-chlorophenyl)-5-[(piperidin-1-yl)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06357393
  • Cas Number : 65924-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09373
Availability details for
Quantity
  • Availability
  • Add to Cart