ENA024295414 Enamine

4-[4-(2-methyl-5-nitrobenzenesulfonyl)piperazin-1-yl]-4-oxobutanoic acid

  • MDL number MFCD05268301
  • Cas Number : 744264-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09378
Availability details for
Quantity
  • Availability
  • Add to Cart