ENA024295416 Enamine

2-chloro-5-(chlorodifluoromethyl)pyridine

  • MDL number MFCD00519716
  • Cas Number : 71701-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09380
Availability details for
Quantity
  • Availability
  • Add to Cart