ENA024295422 Enamine

2-({[(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)carbamoyl]methyl}sulfanyl)benzoic acid

  • MDL number MFCD06015850
  • Cas Number : 630084-03-6
  • Molecular Weight : 397.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-09386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09386
Availability details for
Quantity
  • Availability
  • Add to Cart