ENA024295443 Enamine

5-(2-chlorophenyl)-3-(3-ethoxypropyl)-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD05270859
  • Cas Number : 747411-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09407
Availability details for
Quantity
  • Availability
  • Add to Cart