ENA024295453 Enamine

6-amino-1-methyl-5-piperidin-1-ylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD06356483
  • Cas Number : 790232-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09417

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09417
Availability details for
Quantity
  • Availability
  • Add to Cart