ENA024295457 Enamine

2-[(3-methylphenoxy)methyl]oxirane

  • MDL number MFCD00046990
  • Cas Number : 2186-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09421

Safety Information

signalword Warning
pcodes P201,P202,P261,P264,P272,P273,P280,P281,P302+P352,P308+P313,P332+P313,P333+P313,P362,P363,P391,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-09421
pictograms GHS07,GHS08,GHS09
hstatements H315,H317,H341,H411
Availability details for
Quantity
  • Availability
  • Add to Cart