ENA024295465 Enamine

2-{[(thiophen-2-yl)methoxy]methyl}oxirane

  • MDL number MFCD06655034
  • Cas Number : 80910-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09429
Availability details for
Quantity
  • Availability
  • Add to Cart