ENA024295476 Enamine

3-[(3-methylpiperidin-1-yl)sulfonyl]benzoic acid

  • MDL number MFCD05270853
  • Cas Number : 603118-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09440
Availability details for
Quantity
  • Availability
  • Add to Cart