ENA024295478 Enamine

3-[(isopropylamino)sulfonyl]benzoic acid

  • MDL number MFCD05270857
  • Cas Number : 716358-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09442

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09442
Availability details for
Quantity
  • Availability
  • Add to Cart