ENA024295487 Enamine

4-{[(2,5-dimethylphenyl)sulfonyl]amino}butanoic acid

  • MDL number MFCD05263667
  • Cas Number : 743451-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09451

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09451
Availability details for
Quantity
  • Availability
  • Add to Cart