ENA024295489 Enamine

3-[(dipropylamino)sulfonyl]benzoic acid

  • MDL number MFCD05263650
  • Cas Number : 53212-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09453

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09453
Availability details for
Quantity
  • Availability
  • Add to Cart